The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190964.2 | NP_001177893.1 | 19 - 20 | 3229 | 125 |
NM_001190965.2 | NP_001177894.1 | 18 - 19 | 3244 | 125 | |
NM_003453.4 | NP_003444.1 | 19 - 20 | 3337 | 125 | |
NM_197968.3 | NP_932072.1 | 18 - 19 | 3290 | 125 | |
XM_005266520.3 | XP_005266577.1 | 18 - 19 | 3249 | 125 | |
XM_011535216.2 | XP_011533518.1 | 19 - 20 | 2919 | 125 | |
XM_011535222.1 | XP_011533524.1 | 18 - 19 | 3180 | 125 | |
XM_011535223.2 | XP_011533525.1 | 12 - 13 | 5634 | 125 | |
XM_011535224.2 | XP_011533526.1 | 12 - 13 | 1535 | 125 | |
XM_017020728.1 | XP_016876217.1 | 18 - 19 | 3180 | 125 | |
XM_017020729.1 | XP_016876218.1 | 18 - 19 | 3155 | 125 | |
XM_017020731.1 | XP_016876220.1 | 19 - 20 | 2972 | 125 | |
XM_017020732.1 | XP_016876221.1 | 19 - 20 | 2994 | 125 | |
XM_017020733.1 | XP_016876222.1 | 20 - 21 | 3021 | 125 | |
XM_017020734.1 | XP_016876223.1 | 20 - 21 | 3007 | 125 | |
XM_017020735.1 | XP_016876224.1 | 19 - 20 | 2971 | 125 | |
XM_017020736.1 | XP_016876225.1 | 19 - 20 | 3143 | 125 | |
XM_017020737.1 | XP_016876226.1 | 19 - 20 | 2918 | 125 | |
XM_017020738.1 | XP_016876227.1 | 18 - 19 | 3233 | 125 | |
GenBank mRNA | AF012126.1 | - | 12 - 13 | 1632 | 125 |
AF060181.1 | - | 16 - 17 | 2587 | 125 | |
AJ224901.1 | - | 19 - 20 | 3223 | 125 | |
AK226118.1 | - | 18 - 19 | 3224 | 125 | |
AK289741.1 | - | 18 - 19 | 3215 | 125 | |
BC036372.1 | - | 18 - 19 | 3290 | 125 | |
BX647944.1 | - | 18 - 19 | 3244 | 125 | |
BX648905.1 | - | 11 - 12 | 3674 | 125 | |
Y13472.1 | - | 19 - 20 | 3235 | 125 |