The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145031.2 | NP_001138503.1 | 5 - 6 | 893 | 95 |
NM_001319191.1 | NP_001306120.1 | 5 - 6 | 580 | 95 | |
NM_002658.4 | NP_002649.1 | 6 - 7 | 608 | 95 | |
XM_011539866.2 | XP_011538168.1 | 6 - 7 | 603 | 95 | |
GenBank mRNA | A18397.1 | - | 6 - 7 | 558 | 95 |
A35395.1 | - | 6 - 7 | 528 | 95 | |
AK297940.1 | - | 5 - 6 | 549 | 95 | |
AK298560.1 | - | 5 - 6 | 95 | ||
AK312960.1 | - | 6 - 7 | 577 | 95 | |
BC013575.2 | - | 6 - 7 | 551 | 95 | |
BQ920793.1 | - | 6 - 7 | 601 | 95 | |
BT007391.1 | - | 5 - 6 | 459 | 95 | |
BX360462.2 | - | 6 - 7 | 562 | 95 | |
D00244.1 | - | 6 - 7 | 525 | 95 | |
D11143.1 | - | 4 - 5 | 399 | 95 | |
M15476.1 | - | 6 - 7 | 535 | 95 |