The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300800.1 | NP_001287729.1 | 5 - 6 | 600 | 69 |
NM_024771.3 | NP_079047.2 | 5 - 6 | 529 | 69 | |
XM_011545254.2 | XP_011543556.1 | 6 - 7 | 603 | 69 | |
GenBank mRNA | AK023910.1 | - | 5 - 6 | 498 | 69 |
AK299041.1 | - | 5 - 6 | 594 | 69 | |
AK300451.1 | - | 4 - 5 | 450 | 69 | |
BC041617.1 | - | 5 - 6 | 458 | 69 | |
BC052298.1 | - | 5 - 6 | 458 | 69 | |
BX647309.1 | - | 4 - 5 | 1555 | 69 | |
JF263566.1 | - | 5 - 6 | 498 | 69 |