The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006509.3 | NP_006500.2 | 9 - 10 | 1334 | 72 |
XM_005259127.3 | XP_005259184.1 | 8 - 9 | 1304 | 72 | |
GenBank mRNA | AK290594.1 | - | 9 - 10 | 1241 | 72 |
BC028013.1 | - | 9 - 10 | 1286 | 72 |