The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130955.1 | NP_001124427.1 | 13 - 14 | 2588 | 110 |
NM_015318.3 | NP_056133.2 | 14 - 15 | 2067 | 110 | |
XM_005272464.4 | XP_005272521.1 | 23 - 24 | 3218 | 110 | |
XM_006722705.3 | XP_006722768.1 | 23 - 24 | 3350 | 110 | |
XM_006722706.3 | XP_006722769.1 | 24 - 25 | 3513 | 110 | |
XM_006722708.2 | XP_006722771.1 | 14 - 15 | 2195 | 110 | |
XM_011527835.2 | XP_011526137.1 | 23 - 24 | 3218 | 110 | |
XM_011527836.2 | XP_011526138.1 | 23 - 24 | 3218 | 110 | |
XM_011527837.2 | XP_011526139.1 | 23 - 24 | 3218 | 110 | |
XM_011527838.2 | XP_011526140.1 | 22 - 23 | 2906 | 110 | |
XM_011527839.2 | XP_011526141.1 | 22 - 23 | 2972 | 110 | |
XM_011527840.1 | XP_011526142.1 | 14 - 15 | 2067 | 110 | |
GenBank mRNA | AB011093.1 | - | 14 - 15 | 1968 | 110 |
AK127045.1 | - | 13 - 14 | 2588 | 110 | |
BC077721.1 | - | 15 - 16 | 2135 | 110 |