The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130955.1 | NP_001124427.1 | 16 - 17 | 3172 | 67 |
NM_015318.3 | NP_056133.2 | 17 - 18 | 2651 | 67 | |
XM_005272464.4 | XP_005272521.1 | 26 - 27 | 3802 | 67 | |
XM_006722705.3 | XP_006722768.1 | 26 - 27 | 3934 | 67 | |
XM_006722706.3 | XP_006722769.1 | 27 - 28 | 4097 | 67 | |
XM_006722708.2 | XP_006722771.1 | 17 - 18 | 2779 | 67 | |
XM_011527835.2 | XP_011526137.1 | 26 - 27 | 3802 | 67 | |
XM_011527836.2 | XP_011526138.1 | 26 - 27 | 3802 | 67 | |
XM_011527837.2 | XP_011526139.1 | 26 - 27 | 3802 | 67 | |
XM_011527838.2 | XP_011526140.1 | 25 - 26 | 3490 | 67 | |
XM_011527839.2 | XP_011526141.1 | 25 - 26 | 3556 | 67 | |
XM_011527840.1 | XP_011526142.1 | 17 - 18 | 2651 | 67 | |
GenBank mRNA | AB011093.1 | - | 17 - 18 | 2552 | 67 |
AK074372.1 | - | 12 - 13 | 1834 | 67 | |
AK127045.1 | - | 16 - 17 | 3172 | 67 | |
BC077721.1 | - | 18 - 19 | 2719 | 67 |