The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130955.1 | NP_001124427.1 | 7 - 8 | 1791 | 62 |
NM_015318.3 | NP_056133.2 | 8 - 9 | 1270 | 62 | |
XM_005272464.4 | XP_005272521.1 | 17 - 18 | 2421 | 62 | |
XM_006722705.3 | XP_006722768.1 | 17 - 18 | 2553 | 62 | |
XM_006722706.3 | XP_006722769.1 | 18 - 19 | 2716 | 62 | |
XM_006722708.2 | XP_006722771.1 | 8 - 9 | 1398 | 62 | |
XM_011527835.2 | XP_011526137.1 | 17 - 18 | 2421 | 62 | |
XM_011527836.2 | XP_011526138.1 | 17 - 18 | 2421 | 62 | |
XM_011527837.2 | XP_011526139.1 | 17 - 18 | 2421 | 62 | |
XM_011527838.2 | XP_011526140.1 | 16 - 17 | 2109 | 62 | |
XM_011527839.2 | XP_011526141.1 | 16 - 17 | 2175 | 62 | |
XM_011527840.1 | XP_011526142.1 | 8 - 9 | 1270 | 62 | |
XM_011527841.2 | XP_011526143.1 | 17 - 18 | 2414 | 62 | |
GenBank mRNA | AB011093.1 | - | 8 - 9 | 1171 | 62 |
AK074372.1 | - | 3 - 4 | 471 | 62 | |
AK127045.1 | - | 7 - 8 | 1791 | 62 | |
BC077721.1 | - | 9 - 10 | 1338 | 62 |