The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289177.1 | NP_001276106.1 | 2 - 3 | 388 | 116 |
NM_001289178.1 | NP_001276107.1 | 2 - 3 | 334 | 116 | |
NM_001289179.1 | NP_001276108.1 | 2 - 3 | 379 | 116 | |
NM_001289180.1 | NP_001276109.1 | 3 - 4 | 219 | 116 | |
NM_030924.4 | NP_112186.3 | 2 - 3 | 379 | 116 | |
XM_005259653.3 | XP_005259710.1 | 2 - 3 | 361 | 116 | |
XM_017027332.1 | XP_016882821.1 | 2 - 3 | 206 | 116 | |
XM_017027333.1 | XP_016882822.1 | 2 - 3 | 361 | 116 | |
XM_017027334.1 | XP_016882823.1 | 2 - 3 | 361 | 116 | |
GenBank mRNA | AJ577571.1 | - | 2 - 3 | 282 | 116 |
AK093433.1 | - | 2 - 3 | 347 | 116 | |
AK225862.1 | - | 1 - 2 | 87 | 116 | |
AK302604.1 | - | 2 - 3 | 336 | 116 | |
AL136854.1 | - | 2 - 3 | 120 | 116 | |
AY009107.1 | - | 3 - 4 | 212 | 116 | |
BC022027.1 | - | 2 - 3 | 310 | 116 | |
BC090046.1 | - | 2 - 3 | 295 | 116 | |
BI561348.1 | - | 2 - 3 | 340 | 116 | |
HY011431.1 | - | 2 - 3 | 366 | 116 |