Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPN1
Entrez Gene ID
823
Gene Name
calpain 1
Gene Aliases
CANP, CANP1, CANPL1, SPG76, muCANP, muCL
Chromosome Location
Chr.11: 65181215 - 65212006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198868.1 NP_001185797.1 19 - 20 2142 69
NM_001198869.1 NP_001185798.1 19 - 20 2111 69
NM_005186.3 NP_005177.2 19 - 20 2127 69
NR_040008.1 18 - 19 2057 69
XM_006718698.1 XP_006718761.1 19 - 20 2042 69
XM_011545292.1 XP_011543594.1 19 - 20 2053 69
GenBank mRNA AK025380.1 - 2 - 3 495 69
AK097277.1 - 2 - 3 1522 69
AK301538.1 - 18 - 19 1949 69
AK315556.1 - 19 - 20 2056 69
AY550975.1 - 18 - 19 1950 69
BC008751.2 - 19 - 20 2078 69
BC017200.2 - 19 - 20 1964 69
BC075862.1 - 19 - 20 2045 69
X04366.1 - 19 - 20 2088 69

View Full Product Details