The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002850.3 | NP_002841.3 | 16 - 17 | 2606 | 131 |
XM_005259600.2 | XP_005259657.1 | 13 - 14 | 2535 | 131 | |
XM_017027065.1 | XP_016882554.1 | 14 - 15 | 2547 | 131 | |
XM_017027066.1 | XP_016882555.1 | 14 - 15 | 2565 | 131 | |
XM_017027067.1 | XP_016882556.1 | 14 - 15 | 2463 | 131 | |
XM_017027068.1 | XP_016882557.1 | 14 - 15 | 2547 | 131 | |
XM_017027070.1 | XP_016882559.1 | 14 - 15 | 2547 | 131 | |
XM_017027071.1 | XP_016882560.1 | 14 - 15 | 2547 | 131 | |
XM_017027073.1 | XP_016882562.1 | 8 - 9 | 1750 | 131 | |
GenBank mRNA | U35234.1 | - | 15 - 16 | 2480 | 131 |