The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002850.3 | NP_002841.3 | 18 - 19 | 3339 | 83 |
NM_130854.2 | NP_570924.2 | 14 - 15 | 3273 | 83 | |
XM_005259600.2 | XP_005259657.1 | 15 - 16 | 3268 | 83 | |
XM_005259606.2 | XP_005259663.1 | 14 - 15 | 3241 | 83 | |
XM_005259607.2 | XP_005259664.1 | 14 - 15 | 3241 | 83 | |
XM_011528158.2 | XP_011526460.1 | 3 - 4 | 747 | 83 | |
XM_017027065.1 | XP_016882554.1 | 16 - 17 | 3280 | 83 | |
XM_017027066.1 | XP_016882555.1 | 16 - 17 | 3298 | 83 | |
XM_017027067.1 | XP_016882556.1 | 16 - 17 | 3196 | 83 | |
XM_017027068.1 | XP_016882557.1 | 16 - 17 | 3280 | 83 | |
XM_017027069.1 | XP_016882558.1 | 15 - 16 | 3253 | 83 | |
XM_017027070.1 | XP_016882559.1 | 16 - 17 | 3280 | 83 | |
XM_017027071.1 | XP_016882560.1 | 16 - 17 | 3280 | 83 | |
XM_017027072.1 | XP_016882561.1 | 15 - 16 | 3253 | 83 | |
XM_017027073.1 | XP_016882562.1 | 10 - 11 | 2483 | 83 | |
GenBank mRNA | CN413752.1 | - | 1 - 2 | 519 | 83 |
U35234.1 | - | 17 - 18 | 3213 | 83 | |
U40317.1 | - | 13 - 14 | 3064 | 83 |