Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRS
Entrez Gene ID
5802
Gene Name
protein tyrosine phosphatase, receptor type S
Gene Aliases
PTPSIGMA, R-PTP-S, R-PTP-sigma
Chromosome Location
Chr.19: 5205503 - 5340803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002850.3 NP_002841.3 20 - 21 3687 80
NM_130853.2 NP_570923.2 12 - 13 2394 80
NM_130854.2 NP_570924.2 16 - 17 3621 80
NM_130855.2 NP_570925.2 13 - 14 2406 80
XM_005259600.2 XP_005259657.1 17 - 18 3616 80
XM_005259606.2 XP_005259663.1 16 - 17 3589 80
XM_005259607.2 XP_005259664.1 16 - 17 3589 80
XM_005259609.1 XP_005259666.1 12 - 13 2361 80
XM_005259610.1 XP_005259667.1 12 - 13 2361 80
XM_011528157.2 XP_011526459.1 11 - 12 2232 80
XM_011528158.2 XP_011526460.1 5 - 6 1095 80
XM_017027065.1 XP_016882554.1 18 - 19 3628 80
XM_017027066.1 XP_016882555.1 18 - 19 3646 80
XM_017027067.1 XP_016882556.1 18 - 19 3544 80
XM_017027068.1 XP_016882557.1 18 - 19 3628 80
XM_017027069.1 XP_016882558.1 17 - 18 3601 80
XM_017027070.1 XP_016882559.1 18 - 19 3628 80
XM_017027071.1 XP_016882560.1 18 - 19 3628 80
XM_017027072.1 XP_016882561.1 17 - 18 3601 80
XM_017027073.1 XP_016882562.1 12 - 13 2831 80
XM_017027074.1 XP_016882563.1 13 - 14 2373 80
XM_017027075.1 XP_016882564.1 13 - 14 2373 80
XM_017027076.1 XP_016882565.1 13 - 14 2373 80
GenBank mRNA AB209333.1 - 12 - 13 2394 80
BC104812.1 - 11 - 12 2234 80
BC143286.1 - 11 - 12 2234 80
BC143287.1 - 11 - 12 2234 80
U35234.1 - 19 - 20 3561 80
U40317.1 - 15 - 16 3412 80
U41725.1 - 11 - 12 2185 80

View Full Product Details