The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320687.1 | NP_001307616.1 | 6 - 7 | 1436 | 93 |
NM_016166.2 | NP_057250.1 | 5 - 6 | 837 | 93 | |
XM_011522126.2 | XP_011520428.1 | 7 - 8 | 1544 | 93 | |
XM_011522127.2 | XP_011520429.1 | 5 - 6 | 1321 | 93 | |
XM_017022688.1 | XP_016878177.1 | 5 - 6 | 1315 | 93 | |
XM_017022689.1 | XP_016878178.1 | 5 - 6 | 1328 | 93 | |
XM_017022690.1 | XP_016878179.1 | 5 - 6 | 1133 | 93 | |
XM_017022691.1 | XP_016878180.1 | 5 - 6 | 907 | 93 | |
GenBank mRNA | AF077951.1 | - | 5 - 6 | 689 | 93 |
AF167160.1 | - | 5 - 6 | 785 | 93 | |
AK094641.1 | - | 6 - 7 | 1436 | 93 | |
AK314515.1 | - | 5 - 6 | 712 | 93 | |
AY826819.1 | - | 5 - 6 | 686 | 93 | |
BC053685.1 | - | 5 - 6 | 690 | 93 | |
BC104646.1 | - | 5 - 6 | 705 | 93 | |
BC118587.1 | - | 5 - 6 | 739 | 93 | |
BC121797.1 | - | 5 - 6 | 736 | 93 | |
EF487997.1 | - | 5 - 6 | 689 | 93 | |
EF487998.1 | - | 5 - 6 | 689 | 93 | |
FJ997900.1 | - | 5 - 6 | 689 | 93 | |
U78524.1 | - | 4 - 5 | 671 | 93 |