Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHIPL2
Entrez Gene ID
79802
Gene Name
HHIP like 2
Gene Aliases
KIAA1822L
Chromosome Location
Chr.1: 222501521 - 222548102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024746.3 NP_079022.2 3 - 4 1173 78
XM_011509985.2 XP_011508287.1 3 - 4 1118 78
XM_011509986.1 XP_011508288.1 3 - 4 1118 78
XM_017002350.1 XP_016857839.1 3 - 4 1118 78
GenBank mRNA AK023902.1 - 3 - 4 858 78
AY358602.1 - 3 - 4 1155 78
BC007638.1 - 3 - 4 1159 78
BC015201.1 - 2 - 3 764 78
BC016552.1 - 3 - 4 1173 78

View Full Product Details