The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032880.4 | NP_116269.3 | 5 - 6 | 923 | 61 |
XM_017002604.1 | XP_016858093.1 | 5 - 6 | 569 | 61 | |
XM_017002605.1 | XP_016858094.1 | 4 - 5 | 386 | 61 | |
GenBank mRNA | AK075316.1 | - | 5 - 6 | 923 | 61 |
AK123172.1 | - | 1 - 2 | 864 | 61 | |
AK294121.1 | - | 5 - 6 | 492 | 61 | |
BC007618.1 | - | 5 - 6 | 843 | 61 |