The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172238.3 | NP_758438.2 | 2 - 3 | 1046 | 113 |
GenBank mRNA | AY028376.1 | - | 2 - 3 | 1046 | 113 |