The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287522.1 | NP_001274451.1 | 5 - 6 | 402 | 62 |
NM_001287524.1 | NP_001274453.1 | 2 - 3 | 307 | 62 | |
NM_021872.3 | NP_068658.1 | 5 - 6 | 1254 | 62 | |
GenBank mRNA | BX640836.1 | - | 2 - 3 | 299 | 62 |
DA065196.1 | - | 2 - 3 | 307 | 62 | |
FR695901.1 | - | 5 - 6 | 402 | 62 | |
Z68092.1 | - | 5 - 6 | 562 | 62 |