Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
Chromosome Location
Chr.20: 62952647 - 62968597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302643.1 NP_001289572.1 12 - 13 1359 90
NM_022082.3 NP_071365.3 11 - 12 1297 90
XM_011528978.2 XP_011527280.1 11 - 12 1209 90
GenBank mRNA AK027065.1 - 4 - 5 286 90
AK074107.1 - 10 - 11 2093 90
AK095473.1 - 11 - 12 1148 90
AK125597.1 - 11 - 12 1820 90
AK298503.1 - 11 - 12 1203 90
BC025312.1 - 12 - 13 1299 90
BC027447.1 - 7 - 8 518 90
BC038593.1 - 11 - 12 1139 90

View Full Product Details