Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
Chromosome Location
Chr.20: 62952647 - 62968597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302643.1 NP_001289572.1 13 - 14 1395 75
NM_022082.3 NP_071365.3 12 - 13 1333 75
XM_011528978.2 XP_011527280.1 12 - 13 1245 75
GenBank mRNA AK095473.1 - 12 - 13 1184 75
AK125597.1 - 12 - 13 1856 75
BC025312.1 - 13 - 14 1335 75
BC027447.1 - 8 - 9 554 75
BC038593.1 - 12 - 13 1175 75

View Full Product Details