Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
Chromosome Location
Chr.20: 62952647 - 62968597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302643.1 NP_001289572.1 5 - 6 741 94
NM_022082.3 NP_071365.3 4 - 5 679 94
XM_011528978.2 XP_011527280.1 4 - 5 591 94
GenBank mRNA AK074107.1 - 4 - 5 615 94
AK095473.1 - 4 - 5 530 94
AK125597.1 - 5 - 6 684 94
AK298503.1 - 4 - 5 585 94
BC025312.1 - 5 - 6 681 94
BC038593.1 - 4 - 5 521 94

View Full Product Details