The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000484.3 | NP_000475.1 | 12 - 13 | 1786 | 85 |
NM_001136016.3 | NP_001129488.1 | 11 - 12 | 1710 | 85 | |
NM_001136129.2 | NP_001129601.1 | 9 - 10 | 1393 | 85 | |
NM_001136130.2 | NP_001129602.1 | 11 - 12 | 1618 | 85 | |
NM_001136131.2 | NP_001129603.1 | 10 - 11 | 1448 | 85 | |
NM_001204301.1 | NP_001191230.1 | 12 - 13 | 1786 | 85 | |
NM_001204302.1 | NP_001191231.1 | 11 - 12 | 1729 | 85 | |
NM_001204303.1 | NP_001191232.1 | 10 - 11 | 1561 | 85 | |
NM_201413.2 | NP_958816.1 | 11 - 12 | 1729 | 85 | |
NM_201414.2 | NP_958817.1 | 10 - 11 | 1561 | 85 | |
GenBank mRNA | A02759.1 | - | 10 - 11 | 1507 | 85 |
AF282245.1 | - | 9 - 10 | 1320 | 85 | |
AK294534.1 | - | 11 - 12 | 1565 | 85 | |
AK295373.1 | - | 2 - 3 | 286 | 85 | |
AK295621.1 | - | 11 - 12 | 1671 | 85 | |
AK296229.1 | - | 10 - 11 | 1448 | 85 | |
AK297229.1 | - | 8 - 9 | 924 | 85 | |
AK297412.1 | - | 9 - 10 | 1290 | 85 | |
AK298861.1 | - | 9 - 10 | 1463 | 85 | |
AK312326.1 | - | 12 - 13 | 1651 | 85 | |
BC065529.1 | - | 11 - 12 | 1660 | 85 | |
M16765.1 | - | 5 - 6 | 521 | 85 | |
M18734.1 | - | 1 - 2 | 69 | 85 | |
M28373.1 | - | 9 - 10 | 1292 | 85 | |
X06989.1 | - | 11 - 12 | 1653 | 85 | |
Y00264.1 | - | 10 - 11 | 1508 | 85 |