Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
Chromosome Location
Chr.21: 25880550 - 26171128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000484.3 NP_000475.1 8 - 9 1293 74
NM_001136130.2 NP_001129602.1 7 - 8 1125 74
NM_001204301.1 NP_001191230.1 8 - 9 1293 74

View Full Product Details