Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNPR
Entrez Gene ID
132204
Gene Name
synaptoporin
Gene Aliases
SPO
Chromosome Location
Chr.3: 63200604 - 63616924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144642.4 NP_653243.1 1 - 2 394 97
GenBank mRNA AF411860.2 - 1 - 2 143 97
AK312465.1 - 1 - 2 299 97
AK315958.1 - 1 - 2 357 97
AL834457.1 - 1 - 2 123 97
BC022518.1 - 1 - 2 143 97
DA505318.1 - 1 - 2 395 97

View Full Product Details