The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134778.1 | NP_001128250.1 | 2 - 3 | 305 | 106 |
NM_006195.5 | NP_006186.1 | 2 - 3 | 391 | 106 | |
XM_006717130.1 | XP_006717193.1 | 2 - 3 | 357 | 106 | |
XM_006717131.1 | XP_006717194.1 | 2 - 3 | 357 | 106 | |
XM_006717132.1 | XP_006717195.1 | 2 - 3 | 357 | 106 | |
XM_011518755.1 | XP_011517057.1 | 2 - 3 | 305 | 106 | |
XM_017014786.1 | XP_016870275.1 | 2 - 3 | 305 | 106 | |
GenBank mRNA | AK297093.1 | - | 2 - 3 | 260 | 106 |
AK309713.1 | - | 2 - 3 | 305 | 106 | |
AK312717.1 | - | 2 - 3 | 320 | 106 | |
BC094883.1 | - | 2 - 3 | 342 | 106 | |
X59841.1 | - | 2 - 3 | 387 | 106 |