Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCGBP
Entrez Gene ID
8857
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
FC(GAMMA)BP
Chromosome Location
Chr.19: 39863323 - 39934626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003890.2 NP_003881.2 - 14785 121
GenBank mRNA BM781647.1 - 1 - 2 456 121
D84239.1 - - 14785 121

View Full Product Details