The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277198.1 | NP_001264127.1 | 4 - 5 | 458 | 76 |
NM_001277199.1 | NP_001264128.1 | 4 - 5 | 783 | 76 | |
NM_014184.3 | NP_054903.1 | 4 - 5 | 505 | 76 | |
NR_102347.1 | 3 - 4 | 486 | 76 | ||
GenBank mRNA | AF161512.1 | - | 4 - 5 | 424 | 76 |
AK024569.1 | - | - | 420 | 76 | |
BC000573.1 | - | 4 - 5 | 420 | 76 | |
BI752106.1 | - | 4 - 5 | 374 | 76 | |
DA185135.1 | - | 4 - 5 | 506 | 76 | |
HY018269.1 | - | 3 - 4 | 402 | 76 |