Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIG1
Entrez Gene ID
3978
Gene Name
DNA ligase 1
Gene Aliases
-
Chromosome Location
Chr.19: 48115445 - 48170603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000234.2 NP_000225.1 18 - 19 2144 70
NM_001289063.1 NP_001275992.1 17 - 18 2051 70
NM_001289064.1 NP_001275993.1 16 - 17 1940 70
NM_001320970.1 NP_001307899.1 18 - 19 2141 70
NM_001320971.1 NP_001307900.1 17 - 18 2054 70
NR_110296.1 18 - 19 2141 70
NR_135497.1 18 - 19 2144 70
NR_135498.1 16 - 17 1973 70
NR_135499.1 17 - 18 2054 70
NR_135500.1 18 - 19 2321 70
NR_135501.1 16 - 17 1965 70
XM_017026809.1 XP_016882298.1 16 - 17 1673 70
GenBank mRNA AB208791.1 - 18 - 19 1864 70
AK297296.1 - 16 - 17 1651 70
AK300370.1 - 17 - 18 1792 70
AK300607.1 - 13 - 14 1480 70
AK314210.1 - 18 - 19 1851 70
BC108318.1 - 18 - 19 2144 70
BC110622.2 - 17 - 18 1793 70
M36067.1 - 18 - 19 1844 70

View Full Product Details