The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 11 - 12 | 1600 | 104 |
XM_005253590.3 | XP_005253647.1 | 11 - 12 | 1604 | 104 | |
XM_011538598.2 | XP_011536900.1 | 11 - 12 | 1604 | 104 | |
XM_011538600.2 | XP_011536902.1 | 11 - 12 | 1604 | 104 | |
GenBank mRNA | AB040950.1 | - | 8 - 9 | 972 | 104 |
BC002575.2 | - | 8 - 9 | 983 | 104 | |
BC037964.1 | - | 11 - 12 | 1582 | 104 |