The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 16 - 17 | 2257 | 104 |
XM_005253590.3 | XP_005253647.1 | 16 - 17 | 2261 | 104 | |
XM_011538598.2 | XP_011536900.1 | 16 - 17 | 2261 | 104 | |
XM_011538600.2 | XP_011536902.1 | 16 - 17 | 2261 | 104 | |
GenBank mRNA | AB040950.1 | - | 13 - 14 | 1629 | 104 |
BC002575.2 | - | 13 - 14 | 1640 | 104 | |
BC037964.1 | - | 16 - 17 | 2239 | 104 |