The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 26 - 27 | 3493 | 74 |
GenBank mRNA | AK123968.1 | - | 5 - 6 | 1702 | 74 |
BC002575.2 | - | 23 - 24 | 2876 | 74 | |
BC037964.1 | - | 26 - 27 | 3475 | 74 | |
BC071824.1 | - | 1 - 2 | 84 | 74 |