Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT81
Entrez Gene ID
28981
Gene Name
intraflagellar transport 81
Gene Aliases
CDV-1, CDV-1R, CDV1, CDV1R, DV1
Chromosome Location
Chr.12: 110124335 - 110218795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143779.1 NP_001137251.1 11 - 12 1311 140
NM_014055.3 NP_054774.2 11 - 12 1729 140
XM_017019217.1 XP_016874706.1 11 - 12 1421 140
XM_017019218.1 XP_016874707.1 10 - 11 1565 140
GenBank mRNA AF139540.1 - 11 - 12 1269 140
AF250326.1 - 10 - 11 1222 140
AF332010.1 - 11 - 12 1687 140
AK000874.1 - 4 - 5 491 140
AK091549.1 - 10 - 11 1089 140
AK291043.1 - 11 - 12 1631 140

View Full Product Details