Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT81
Entrez Gene ID
28981
Gene Name
intraflagellar transport 81
Gene Aliases
CDV-1, CDV-1R, CDV1, CDV1R, DV1
Chromosome Location
Chr.12: 110124335 - 110218795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143779.1 NP_001137251.1 14 - 15 1685 79
NM_014055.3 NP_054774.2 14 - 15 2103 79
XM_017019217.1 XP_016874706.1 14 - 15 1795 79
XM_017019218.1 XP_016874707.1 13 - 14 1939 79
GenBank mRNA AF139540.1 - 14 - 15 1643 79
AF250326.1 - 13 - 14 1596 79
AF332010.1 - 14 - 15 2061 79
AK000874.1 - 7 - 8 866 79
AK091549.1 - 13 - 14 1463 79
AK291043.1 - 14 - 15 2005 79

View Full Product Details