Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT81
Entrez Gene ID
28981
Gene Name
intraflagellar transport 81
Gene Aliases
CDV-1, CDV-1R, CDV1, CDV1R, DV1
Chromosome Location
Chr.12: 110124335 - 110218795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143779.1 NP_001137251.1 7 - 8 824 95
NM_014055.3 NP_054774.2 7 - 8 1242 95
NM_031473.3 NP_113661.2 7 - 8 1242 95
XM_011538232.2 XP_011536534.1 7 - 8 770 95
XM_017019217.1 XP_016874706.1 7 - 8 934 95
XM_017019218.1 XP_016874707.1 7 - 8 1242 95
GenBank mRNA AF139540.1 - 7 - 8 782 95
AF250326.1 - 6 - 7 735 95
AF332010.1 - 7 - 8 1200 95
AK091549.1 - 6 - 7 602 95
AK291043.1 - 7 - 8 1144 95
BC004536.2 - 7 - 8 1078 95
BC005396.1 - 7 - 8 744 95
BC108257.1 - 6 - 7 707 95

View Full Product Details