Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
Chromosome Location
Chr.22: 19966491 - 20016802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001670.2 NP_001661.1 14 - 15 2709 124
XM_005261242.2 XP_005261299.1 13 - 14 2707 124
XM_005261243.3 XP_005261300.1 13 - 14 2707 124
XM_005261244.3 XP_005261301.1 13 - 14 2707 124
XM_006724243.2 XP_006724306.1 14 - 15 2725 124
XM_006724245.2 XP_006724308.1 14 - 15 2725 124
XM_006724246.3 XP_006724309.1 12 - 13 5561 124
XM_006724247.3 XP_006724310.1 12 - 13 2369 124
XM_006724248.3 XP_006724311.1 11 - 12 2350 124
XM_011530179.2 XP_011528481.1 13 - 14 2692 124
XM_011530180.1 XP_011528482.1 14 - 15 2725 124
XM_011530182.2 XP_011528484.1 9 - 10 1337 124
XM_011530183.2 XP_011528485.1 7 - 8 1689 124
GenBank mRNA AK092664.1 - 7 - 8 1156 124
BC137436.1 - 13 - 14 2487 124
U51269.1 - 14 - 15 2710 124

View Full Product Details