Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRRAP
Entrez Gene ID
8295
Gene Name
transformation/transcription domain associated protein
Gene Aliases
PAF350/400, PAF400, STAF40, TR-AP, Tra1
Chromosome Location
Chr.7: 98878490 - 99013243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244580.1 NP_001231509.1 7 - 8 712 106
NM_003496.3 NP_003487.1 7 - 8 712 106
GenBank mRNA AF076974.1 - 7 - 8 712 106
AF110377.1 - 7 - 8 632 106

View Full Product Details