The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005458.7 | NP_005449.5 | 2 - 3 | 921 | 63 |
GenBank mRNA | AF056085.1 | - | 2 - 3 | 918 | 63 |
AF069755.1 | - | 2 - 3 | 617 | 63 | |
AF074483.1 | - | 2 - 3 | 461 | 63 | |
AF095784.1 | - | 2 - 3 | 592 | 63 | |
AF099033.1 | - | 2 - 3 | 461 | 63 | |
AJ012188.1 | - | 2 - 3 | 461 | 63 | |
AK308139.1 | - | 1 - 2 | 94 | 63 | |
BC035071.2 | - | 3 - 4 | 756 | 63 | |
JQ308173.1 | - | 2 - 3 | 461 | 63 |