The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136193.1 | NP_001129665.1 | 5 - 6 | 1523 | 133 |
NM_001136194.1 | NP_001129666.1 | 5 - 6 | 1538 | 133 | |
NM_014929.3 | NP_055744.2 | 5 - 6 | 1430 | 133 | |
GenBank mRNA | AB023188.1 | - | 4 - 5 | 1171 | 133 |
AF223470.1 | - | 5 - 6 | 1297 | 133 | |
AK001516.1 | - | 5 - 6 | 1210 | 133 | |
AK021557.1 | - | 5 - 6 | 1324 | 133 | |
AL833877.1 | - | 5 - 6 | 1204 | 133 | |
BC001544.1 | - | 4 - 5 | 1170 | 133 |