The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006445.3 | NP_006436.3 | 15 - 16 | 2301 | 87 |
GenBank mRNA | AB007510.1 | - | 15 - 16 | 2228 | 87 |
AF092565.1 | - | 15 - 16 | 2225 | 87 | |
BC064370.1 | - | 15 - 16 | 2266 | 87 |