The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006445.3 | NP_006436.3 | 23 - 24 | 3768 | 86 |
GenBank mRNA | AB007510.1 | - | 23 - 24 | 3695 | 86 |
AF092565.1 | - | 23 - 24 | 3692 | 86 | |
BC064370.1 | - | 23 - 24 | 3733 | 86 |