The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000283.3 | NP_000274.2 | 17 - 18 | 2181 | 79 |
NM_001145291.1 | NP_001138763.1 | 17 - 18 | 2181 | 79 | |
NM_001145292.1 | NP_001138764.1 | 15 - 16 | 1560 | 79 | |
XM_011513473.2 | XP_011511775.1 | 18 - 19 | 3448 | 79 | |
XM_011513474.2 | XP_011511776.1 | 18 - 19 | 3448 | 79 | |
XM_011513475.2 | XP_011511777.1 | 20 - 21 | 3551 | 79 | |
XM_011513476.2 | XP_011511778.1 | 18 - 19 | 3448 | 79 | |
XM_011513477.2 | XP_011511779.1 | 13 - 14 | 2300 | 79 | |
XM_011513478.2 | XP_011511780.1 | 14 - 15 | 2367 | 79 | |
XM_017008283.1 | XP_016863772.1 | 15 - 16 | 1587 | 79 | |
XM_017008284.1 | XP_016863773.1 | 15 - 16 | 1542 | 79 | |
XM_017008285.1 | XP_016863774.1 | 15 - 16 | 1713 | 79 | |
XM_017008286.1 | XP_016863775.1 | 15 - 16 | 1542 | 79 | |
XM_017008287.1 | XP_016863776.1 | 14 - 15 | 1643 | 79 | |
XM_017008288.1 | XP_016863777.1 | 12 - 13 | 2252 | 79 | |
XM_017008289.1 | XP_016863778.1 | 13 - 14 | 1440 | 79 | |
GenBank mRNA | AK295285.1 | - | 15 - 16 | 1557 | 79 |
AK316054.1 | - | 15 - 16 | 1517 | 79 | |
BC000249.1 | - | 17 - 18 | 2171 | 79 | |
BT009794.1 | - | 17 - 18 | 2128 | 79 | |
S41458.1 | - | 17 - 18 | 2149 | 79 | |
X66142.1 | - | 17 - 18 | 2133 | 79 |