The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000283.3 | NP_000274.2 | 19 - 20 | 2317 | 111 |
NM_001145291.1 | NP_001138763.1 | 19 - 20 | 2317 | 111 | |
NM_001145292.1 | NP_001138764.1 | 17 - 18 | 1696 | 111 | |
XM_011513473.2 | XP_011511775.1 | 20 - 21 | 3584 | 111 | |
XM_011513474.2 | XP_011511776.1 | 20 - 21 | 3584 | 111 | |
XM_011513475.2 | XP_011511777.1 | 22 - 23 | 3687 | 111 | |
XM_011513476.2 | XP_011511778.1 | 20 - 21 | 3584 | 111 | |
XM_011513477.2 | XP_011511779.1 | 15 - 16 | 2436 | 111 | |
XM_011513478.2 | XP_011511780.1 | 16 - 17 | 2503 | 111 | |
XM_017008283.1 | XP_016863772.1 | 17 - 18 | 1723 | 111 | |
XM_017008284.1 | XP_016863773.1 | 17 - 18 | 1678 | 111 | |
XM_017008285.1 | XP_016863774.1 | 17 - 18 | 1849 | 111 | |
XM_017008286.1 | XP_016863775.1 | 17 - 18 | 1678 | 111 | |
XM_017008287.1 | XP_016863776.1 | 16 - 17 | 1779 | 111 | |
XM_017008288.1 | XP_016863777.1 | 14 - 15 | 2388 | 111 | |
XM_017008289.1 | XP_016863778.1 | 15 - 16 | 1576 | 111 | |
GenBank mRNA | AK295285.1 | - | 17 - 18 | 1693 | 111 |
AK316054.1 | - | 17 - 18 | 1653 | 111 | |
BC000249.1 | - | 19 - 20 | 2307 | 111 | |
BT009794.1 | - | 19 - 20 | 2264 | 111 | |
S41458.1 | - | 19 - 20 | 2285 | 111 | |
X66142.1 | - | 19 - 20 | 2269 | 111 |