Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTBP1
Entrez Gene ID
4052
Gene Name
latent transforming growth factor beta binding protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 32947129 - 33399509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000627.3 NP_000618.3 2 - 3 583 95
NM_001166264.1 NP_001159736.1 2 - 3 583 95
NM_001166265.1 NP_001159737.1 2 - 3 583 95
NM_001166266.1 NP_001159738.1 2 - 3 583 95
NM_206943.2 NP_996826.2 6 - 7 1443 95
XM_005264317.3 XP_005264374.1 6 - 7 1445 95
XM_005264318.3 XP_005264375.1 6 - 7 1445 95
XM_011532853.2 XP_011531155.1 6 - 7 1616 95
XM_011532855.2 XP_011531157.1 6 - 7 1445 95
XM_011532856.2 XP_011531158.1 6 - 7 1445 95
XM_011532857.2 XP_011531159.1 6 - 7 1445 95
XM_011532858.2 XP_011531160.1 6 - 7 1445 95
XM_011532859.2 XP_011531161.1 6 - 7 1445 95
XM_011532860.2 XP_011531162.1 5 - 6 1277 95
XM_011532861.2 XP_011531163.1 6 - 7 1445 95
XM_011532862.2 XP_011531164.1 6 - 7 1445 95
XM_017004108.1 XP_016859597.1 6 - 7 1445 95
XM_017004109.1 XP_016859598.1 6 - 7 1445 95
XM_017004110.1 XP_016859599.1 6 - 7 1445 95
GenBank mRNA AB208801.1 - 2 - 3 541 95
AF489528.1 - 2 - 3 442 95
BC130289.1 - 2 - 3 523 95
BC144127.1 - 2 - 3 523 95
BC144128.1 - 2 - 3 523 95
M34057.1 - 2 - 3 532 95

View Full Product Details