The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004939.2 | NP_004930.1 | 10 - 11 | 938 | 142 |
GenBank mRNA | AB593031.1 | - | 10 - 11 | 691 | 142 |
AB593032.1 | - | 10 - 11 | 668 | 142 | |
AB593033.1 | - | 6 - 7 | 465 | 142 | |
AB593034.1 | - | 10 - 11 | 711 | 142 | |
AK297432.1 | - | 9 - 10 | 569 | 142 | |
AK298212.1 | - | 7 - 8 | 522 | 142 | |
AK298426.1 | - | 10 - 11 | 630 | 142 | |
AK313933.1 | - | 10 - 11 | 657 | 142 | |
BC012132.1 | - | 10 - 11 | 674 | 142 | |
BC053673.1 | - | 10 - 11 | 675 | 142 | |
EF418570.1 | - | 10 - 11 | 643 | 142 | |
EF418571.1 | - | 10 - 11 | 643 | 142 | |
X70649.1 | - | 10 - 11 | 913 | 142 |