Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCMF1
Entrez Gene ID
56888
Gene Name
potassium channel modulatory factor 1
Gene Aliases
DEBT91, FIGC, PCMF, ZZZ1
Chromosome Location
Chr.2: 84971108 - 85059472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006712052.3 XP_006712115.1 1 - 2 392 78

View Full Product Details