The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024940.7 | NP_079216.4 | 28 - 29 | 3129 | 86 |
XM_011544661.1 | XP_011542963.1 | 28 - 29 | 3087 | 86 | |
XM_011544662.1 | XP_011542964.1 | 28 - 29 | 3087 | 86 | |
XM_011544663.2 | XP_011542965.1 | 28 - 29 | 3087 | 86 | |
XM_017013865.1 | XP_016869354.1 | 28 - 29 | 2939 | 86 | |
XM_017013866.1 | XP_016869355.1 | 26 - 27 | 4044 | 86 | |
XM_017013867.1 | XP_016869356.1 | 21 - 22 | 2477 | 86 | |
XM_017013868.1 | XP_016869357.1 | 22 - 23 | 2400 | 86 | |
GenBank mRNA | AK123607.1 | - | 28 - 29 | 3062 | 86 |
AK126249.1 | - | 3 - 4 | 248 | 86 | |
BC137175.1 | - | 28 - 29 | 2987 | 86 | |
BC137176.1 | - | 28 - 29 | 2987 | 86 | |
BX648352.1 | - | 11 - 12 | 1121 | 86 | |
CR749353.1 | - | 20 - 21 | 2270 | 86 | |
CR933718.1 | - | 16 - 17 | 1703 | 86 |