The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024940.7 | NP_079216.4 | 34 - 35 | 3708 | 71 |
XM_011544661.1 | XP_011542963.1 | 34 - 35 | 3666 | 71 | |
XM_011544662.1 | XP_011542964.1 | 34 - 35 | 3666 | 71 | |
XM_011544663.2 | XP_011542965.1 | 34 - 35 | 3666 | 71 | |
XM_017013865.1 | XP_016869354.1 | 34 - 35 | 3518 | 71 | |
XM_017013866.1 | XP_016869355.1 | 32 - 33 | 4623 | 71 | |
XM_017013867.1 | XP_016869356.1 | 27 - 28 | 3056 | 71 | |
XM_017013868.1 | XP_016869357.1 | 28 - 29 | 2979 | 71 | |
GenBank mRNA | AK123607.1 | - | 34 - 35 | 3641 | 71 |
AK126249.1 | - | 9 - 10 | 827 | 71 | |
BC041761.2 | - | 3 - 4 | 289 | 71 | |
BC137175.1 | - | 34 - 35 | 3566 | 71 | |
BC137176.1 | - | 34 - 35 | 3566 | 71 | |
BX648352.1 | - | 17 - 18 | 1662 | 71 | |
CR749353.1 | - | 26 - 27 | 2849 | 71 | |
CR933718.1 | - | 22 - 23 | 2282 | 71 |