The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024940.7 | NP_079216.4 | 8 - 9 | 947 | 91 |
XM_011544661.1 | XP_011542963.1 | 8 - 9 | 884 | 91 | |
XM_011544662.1 | XP_011542964.1 | 8 - 9 | 884 | 91 | |
XM_011544663.2 | XP_011542965.1 | 8 - 9 | 884 | 91 | |
XM_017013865.1 | XP_016869354.1 | 8 - 9 | 736 | 91 | |
XM_017013866.1 | XP_016869355.1 | 6 - 7 | 1841 | 91 | |
XM_017013868.1 | XP_016869357.1 | 2 - 3 | 222 | 91 | |
GenBank mRNA | AK123607.1 | - | 8 - 9 | 884 | 91 |
BC011877.2 | - | 8 - 9 | 898 | 91 | |
BC137175.1 | - | 8 - 9 | 805 | 91 | |
BC137176.1 | - | 8 - 9 | 805 | 91 |