The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000095.2 | NP_000086.2 | 5 - 6 | 564 | 146 |
GenBank mRNA | AB086984.1 | - | 5 - 6 | 528 | 146 |
AK223216.1 | - | 5 - 6 | 564 | 146 | |
AK290595.1 | - | 5 - 6 | 564 | 146 | |
AK297798.1 | - | 4 - 5 | 466 | 146 | |
AK297814.1 | - | 4 - 5 | 525 | 146 | |
AU120397.1 | - | 5 - 6 | 564 | 146 | |
BC033676.1 | - | 4 - 5 | 788 | 146 | |
BC110847.1 | - | 5 - 6 | 550 | 146 | |
BC125092.1 | - | 5 - 6 | 557 | 146 | |
L32137.1 | - | 5 - 6 | 553 | 146 |