The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130955.1 | NP_001124427.1 | 19 - 20 | 3786 | 88 |
NM_015318.3 | NP_056133.2 | 20 - 21 | 3265 | 88 | |
XM_005272464.4 | XP_005272521.1 | 29 - 30 | 4416 | 88 | |
GenBank mRNA | AB011093.1 | - | 20 - 21 | 3166 | 88 |
AK127045.1 | - | 19 - 20 | 3786 | 88 |