The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007527.1 | NP_001007528.1 | 10 - 11 | 1723 | 152 |
XM_011514162.2 | XP_011512464.1 | 10 - 11 | 2165 | 152 | |
XM_017010024.1 | XP_016865513.1 | 11 - 12 | 1841 | 152 | |
GenBank mRNA | AK091295.1 | - | 10 - 11 | 1663 | 152 |
AK300433.1 | - | 11 - 12 | 1633 | 152 | |
AK307575.1 | - | 10 - 11 | 1669 | 152 | |
BC110506.2 | - | 10 - 11 | 1662 | 152 | |
BX647340.1 | - | 10 - 11 | 1698 | 152 | |
CR749399.1 | - | 10 - 11 | 1724 | 153 |